Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3R,5R,6S)-Atogepant | 1374248-81-3 | 99.6% | 603.52 | 100 MG
SDP

Catalog No. 5000346496
Encompass_Preferred

(3R,5R,6S)-Atogepant is the enantiomer of Atogepant, a calcitonin gene-related peptide receptor (CGRP) antagonist. It is intended for research use only, specifically for studying migraine.

  • Enantiomer of Atogepant
  • CGRP receptor antagonist
  • Molecular weight: 603.52
  • Appearance: solid, white to off-white
  • Suitable for migraine research

Catalog No. 50-003-46496 Supplier Medchemexpress LLC Supplier No. HY109022A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(3R,5R,6S)-Atogepant is the enantiomer of Atogepant, a calcitonin gene-related peptide receptor (CGRP) antagonist. It is intended for research use only, specifically for studying migraine.

  • Enantiomer of Atogepant
  • CGRP receptor antagonist
  • Molecular weight: 603.52
  • Appearance: solid, white to off-white
  • Suitable for migraine research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.