Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3R,5R,6S)-Atogepant | 99.55% | 603.52 | 50 MG
SDP

Catalog No. 5000346630
Encompass_Preferred

(3R,5R,6S)-Atogepant, also known as (3R,5R,6S)-MK-8031, is an enantiomer of Atogepant. This compound acts as a calcitonin gene-related peptide receptor (CGRP) antagonist and is utilized in researching migraine.

  • Enantiomer of Atogepant
  • Calcitonin gene-related peptide receptor (CGRP) antagonist
  • Used for researching migraine
  • High purity of 99.55%
  • Molecular weight: 603.52
  • Store powder at -20°C for up to 3 years
  • Store in solvent at -80°C for up to 6 months or -20°C for 1 month
  • Soluble in DMSO (50 mg/mL)

Catalog No. 50-003-46630 Supplier Medchemexpress LLC Supplier No. HY109022A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(3R,5R,6S)-Atogepant, also known as (3R,5R,6S)-MK-8031, is an enantiomer of Atogepant. This compound acts as a calcitonin gene-related peptide receptor (CGRP) antagonist and is utilized in researching migraine.

  • Enantiomer of Atogepant
  • Calcitonin gene-related peptide receptor (CGRP) antagonist
  • Used for researching migraine
  • High purity of 99.55%
  • Molecular weight: 603.52
  • Store powder at -20°C for up to 3 years
  • Store in solvent at -80°C for up to 6 months or -20°C for 1 month
  • Soluble in DMSO (50 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.