Learn More
Cayman Chemical (3R,8S)-Falcarindiol, Purity: ≥98%, CAS: 225110-25-8, 10 mg
SDP
Catalog No. 502978860
Encompass_Preferred
A polyacetylene with diverse biological activities; active against M. tuberculosis strain H37Ra (MIC = 6.25 μg/ml); cytotoxic to MCF-7, H1299, and HepG2 cells (IC50s = 29.4, 22.1, and 23.6 μM, respectively); has algicidal activity against H. circularisquama at 2 and 4 μM
Catalog No. 50-297-8860 Supplier Cayman Chemical Supplier No. 3091910 MG
A polyacetylene with diverse biological activities; active against M. tuberculosis strain H37Ra (MIC = 6.25 μg/ml); cytotoxic to MCF-7, H1299, and HepG2 cells (IC50s = 29.4, 22.1, and 23.6 μM, respectively); has algicidal activity against H. circularisquama at 2 and 4 μM