Learn More
Medchemexpress LLC (3R)-N,N-dimethyl-1-(5-phenylpyrazolo[1,5-a]pyrimidin-7-yl)pyrrolidin-3-amine | 2089389-15-9 | 99.8% | 307.39 g/mol | C18H21N5 | 1 ML
SDP
Catalog No. 5000228683
Encompass_Preferred
(R)-ZINC-3573 is a small-molecule probe and selective agonist of the Mas-related G protein-coupled receptor X2 (MRGPRX2), supplied for research use. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available as solid quantities for assay development and pharmacology studies.
  • Selective MRGPRX2 agonist for pain and itch research.
  • Supplied as 10 mM solution in DMSO, 1 mL volume.
  • High purity suitable for biochemical and cellular assays.
  • Chemical formula C18H21N5; molecular weight 307.39 g/mol.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-28683 Supplier Medchemexpress LLC Supplier No. HY11806910MM 1ML
(R)-ZINC-3573 is a small-molecule probe and selective agonist of the Mas-related G protein-coupled receptor X2 (MRGPRX2), supplied for research use. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available as solid quantities for assay development and pharmacology studies.
  • Selective MRGPRX2 agonist for pain and itch research.
  • Supplied as 10 mM solution in DMSO, 1 mL volume.
  • High purity suitable for biochemical and cellular assays.
  • Chemical formula C18H21N5; molecular weight 307.39 g/mol.
  • For research use only; not for human or diagnostic use.