Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3S)-3-(((2S)-2-((4-methoxy-1H-indole-2-carbonyl)amino)-4-methylpentanoyl)amino)-2-oxo-4-((3S)-... | 870153-29-0 | 99.3% | C24H32N4O6 | 10MG
SDP

Catalog No. 5000261273
Encompass_Preferred

A research-grade small-molecule inhibitor of coronavirus 3C-like protease (3CLpro/Mpro) with potent biochemical activity; used as an active metabolite in antiviral research and as a reference compound for protease-inhibitor studies.

  • Potent inhibition of SARS-CoV-2 3CLpro (IC50 0.27 nM) and SARS-CoV-1 3CLpro (IC50 4 nM).
  • Used as active metabolite and reference standard in antiviral research.
  • Molecular formula C24H32N4O6; molecular weight 472.53 g·mol-1.
  • Supplied as a solid with high purity (99.31%).
  • Storage: powder at -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Suitable for biochemical assays and preclinical studies.

Catalog No. 50-002-61273 Supplier Medchemexpress LLC Supplier No. HY13704810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A research-grade small-molecule inhibitor of coronavirus 3C-like protease (3CLpro/Mpro) with potent biochemical activity; used as an active metabolite in antiviral research and as a reference compound for protease-inhibitor studies.

  • Potent inhibition of SARS-CoV-2 3CLpro (IC50 0.27 nM) and SARS-CoV-1 3CLpro (IC50 4 nM).
  • Used as active metabolite and reference standard in antiviral research.
  • Molecular formula C24H32N4O6; molecular weight 472.53 g·mol-1.
  • Supplied as a solid with high purity (99.31%).
  • Storage: powder at -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Suitable for biochemical assays and preclinical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.