Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3S)-(-)-3-(Ethylamino)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. E04341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04341G Supplier TCI America Supplier No. E04341G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 381670-31-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00191359
InChI Key OPCPWFHLFKAUEA-UHFFFAOYNA-N
PubChem CID 22831501
IUPAC Name N-ethylpyrrolidin-3-amine
SMILES CCNC1CCNC1

Specifications

Color Yellow
Boiling Point 176°C
Quantity 1 g
UN Number 2734
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (3S)-(-)-3-(Ethylamino)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.