Learn More
Medchemexpress LLC (3S,4R,5R)-3,4,5,6-tetrahydroxy-1-[(4-nitro-2,1,3-benzoxadiazol-7-yl)amino]hexan-2-one | 940961-04-6 | 99.7% | 342.26 g/mol | C12H14N4O8 | 1MG
SDP
Catalog No. 5000235750
Encompass_Preferred
NBD-Fructose is a fluorescent derivative of D-fructose produced by coupling a nitrobenzoxadiazole (NBD) group to amino-fructose. It is used as a fluorescent probe in biochemical and cell-based assays to monitor fructose uptake and localization, with excitation and emission suitable for standard fluorescence detectors. Supplied as a powder for research use only.
Catalog No. 50-002-35750 Supplier Medchemexpress LLC Supplier No. HY1230671MG
NBD-Fructose is a fluorescent derivative of D-fructose produced by coupling a nitrobenzoxadiazole (NBD) group to amino-fructose. It is used as a fluorescent probe in biochemical and cell-based assays to monitor fructose uptake and localization, with excitation and emission suitable for standard fluorescence detectors. Supplied as a powder for research use only.