Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3S,4S)-A2-32-01 | 98.4% | 301.42 | 100 MG
SDP

Catalog No. 5000344731
Encompass_Preferred

(3S,4S)-A2-32-01 (compound 24(S,S)) is identified as a less active (S,S)-enantiomer of A2-32-01. A2-32-01 is a potent inhibitor of caseinolytic protein proteases (ClpP). This product is for research use only and is not sold to patients.

  • Target is ClpP
  • Pathway is Cell Cycle/DNA Damage
  • Stored at -20°C, protect from light
  • In solvent stored at -80°C for 6 months or -20°C for 1 month (protect from light)
  • Soluble in DMSO (100 mg/mL; needs ultrasonic)
  • Soluble in various in vivo dissolution protocols

Catalog No. 50-003-44731 Supplier Medchemexpress LLC Supplier No. HY111532B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(3S,4S)-A2-32-01 (compound 24(S,S)) is identified as a less active (S,S)-enantiomer of A2-32-01. A2-32-01 is a potent inhibitor of caseinolytic protein proteases (ClpP). This product is for research use only and is not sold to patients.

  • Target is ClpP
  • Pathway is Cell Cycle/DNA Damage
  • Stored at -20°C, protect from light
  • In solvent stored at -80°C for 6 months or -20°C for 1 month (protect from light)
  • Soluble in DMSO (100 mg/mL; needs ultrasonic)
  • Soluble in various in vivo dissolution protocols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.