Learn More
Medchemexpress LLC (3S,4S)-PF-06459988 | 1858291-14-1 | 98.97% | 431.88 | 5 MG
SDP
Catalog No. 5000403613
Encompass_Preferred
  • (3S, 4S)-PF-06459988 is the S enantiomer of PF-06459988 with less activity.
  • It is a potent irreversible inhibitor of T790M mutant epidermal growth factor receptor (EGFR).
  • Possesses excellent selectivity against EGFR wild-type.
  • Features a minimally reactive electrophile that reduces the propensity of off-target labeling.
Catalog No. 50-004-03613 Supplier Medchemexpress LLC Supplier No. HY19985A5MG
  • (3S, 4S)-PF-06459988 is the S enantiomer of PF-06459988 with less activity.
  • It is a potent irreversible inhibitor of T790M mutant epidermal growth factor receptor (EGFR).
  • Possesses excellent selectivity against EGFR wild-type.
  • Features a minimally reactive electrophile that reduces the propensity of off-target labeling.