Learn More
Medchemexpress LLC (3S,4S)-PF-06459988 | 1858291-14-1 | 98.97% | 431.88 | 50 MG
SDP
Catalog No. 5000404283
Encompass_Preferred
(3S, 4S)-PF-06459988 is the S enantiomer of PF-06459988 with less activity. PF-06459988 is a potent irreversible inhibitor of T790M mutant epidermal growth factor receptor (EGFR). It exhibits excellent selectivity against EGFR wild-type, possessing a minimally reactive electrophile that reduces the propensity of off-target labeling.
  • Potent irreversible inhibitor of T790M mutant EGFR.
  • Excellent selectivity against EGFR wild-type.
  • Minimally reactive electrophile to reduce off-target labeling.
  • For research use only.
Catalog No. 50-004-04283 Supplier Medchemexpress LLC Supplier No. HY19985A50MG
(3S, 4S)-PF-06459988 is the S enantiomer of PF-06459988 with less activity. PF-06459988 is a potent irreversible inhibitor of T790M mutant epidermal growth factor receptor (EGFR). It exhibits excellent selectivity against EGFR wild-type, possessing a minimally reactive electrophile that reduces the propensity of off-target labeling.
  • Potent irreversible inhibitor of T790M mutant EGFR.
  • Excellent selectivity against EGFR wild-type.
  • Minimally reactive electrophile to reduce off-target labeling.
  • For research use only.