Learn More
Medchemexpress LLC (3S,4S)-PF-06459988 | 1858291-14-1 | 99.0% | 10 MG
SDP
Catalog No. 5000297249
Encompass_Preferred
(3S,4S)-PF-06459988 is the S-enantiomer of PF-06459988, provided as a high-purity research compound that irreversibly inhibits the T790M mutant form of the epidermal growth factor receptor (EGFR). It is intended for biochemical and cellular studies of mutant EGFR signaling, inhibitor selectivity, and related preclinical research applications.
  • Has selectivity for T790M mutant EGFR.
  • Acts as an irreversible EGFR inhibitor.
  • Supplied as a high-purity powder suitable for research use.
  • Soluble in DMSO at high concentration; in vivo formulations reported.
  • Stable under recommended frozen storage; follow solvent storage guidelines for prepared solutions.
  • Suitable for biochemical assays, cellular studies, and selectivity profiling.
Catalog No. 50-002-97249 Supplier Medchemexpress LLC Supplier No. HY19985A10MG
(3S,4S)-PF-06459988 is the S-enantiomer of PF-06459988, provided as a high-purity research compound that irreversibly inhibits the T790M mutant form of the epidermal growth factor receptor (EGFR). It is intended for biochemical and cellular studies of mutant EGFR signaling, inhibitor selectivity, and related preclinical research applications.
  • Has selectivity for T790M mutant EGFR.
  • Acts as an irreversible EGFR inhibitor.
  • Supplied as a high-purity powder suitable for research use.
  • Soluble in DMSO at high concentration; in vivo formulations reported.
  • Stable under recommended frozen storage; follow solvent storage guidelines for prepared solutions.
  • Suitable for biochemical assays, cellular studies, and selectivity profiling.