Learn More
Medchemexpress LLC (3S,4S)-PF-06459988 | 1858291-14-1 | 99.0% | 431.88 | 10 MM * 1 ML
SDP
Catalog No. 5000401112
Encompass_Preferred
(3S,4S)-PF-06459988 is the S enantiomer of PF-06459988, exhibiting less activity. PF-06459988 is a potent, irreversible inhibitor of the T790M mutant epidermal growth factor receptor (EGFR). It demonstrates excellent selectivity against wild-type EGFR and contains a minimally reactive electrophile to reduce off-target labeling.
  • Less active S enantiomer of PF-06459988.
  • Irreversible inhibitor of T790M mutant EGFR.
  • Excellent selectivity against EGFR wild-type.
  • Minimally reactive electrophile, reducing off-target labeling.
  • For research use only.
Catalog No. 50-004-01112 Supplier Medchemexpress LLC Supplier No. HY19985A10MM/1ML
(3S,4S)-PF-06459988 is the S enantiomer of PF-06459988, exhibiting less activity. PF-06459988 is a potent, irreversible inhibitor of the T790M mutant epidermal growth factor receptor (EGFR). It demonstrates excellent selectivity against wild-type EGFR and contains a minimally reactive electrophile to reduce off-target labeling.
  • Less active S enantiomer of PF-06459988.
  • Irreversible inhibitor of T790M mutant EGFR.
  • Excellent selectivity against EGFR wild-type.
  • Minimally reactive electrophile, reducing off-target labeling.
  • For research use only.