Learn More
Medchemexpress LLC (3S,4S)-PF-06459988 | 1858291-14-1 | 99.0% | 431.88 | 100 MG
SDP
Catalog No. 5000404295
Encompass_Preferred
(3S, 4S)-PF-06459988 is the S enantiomer of PF-06459988 with less activity. PF-06459988 is a potent irreversible inhibitor of T790M mutant epidermal growth factor receptor (EGFR). It exhibits excellent selectivity against EGFR wild-type and has a minimally reactive electrophile to reduce off-target labeling. This compound is for research use only and not sold to patients.
  • Inhibition of EGF-stimulated wild type EGFR autophosphorylation expressed in human A549 cells
  • Inhibition of EGFR L858R/T790M double mutant autophosphorylation in human NCI-H1975 cells
  • Excellent selectivity against EGFR wild-type
  • Minimally reactive electrophile reduces off-target labeling
Catalog No. 50-004-04295 Supplier Medchemexpress LLC Supplier No. HY19985A100MG
(3S, 4S)-PF-06459988 is the S enantiomer of PF-06459988 with less activity. PF-06459988 is a potent irreversible inhibitor of T790M mutant epidermal growth factor receptor (EGFR). It exhibits excellent selectivity against EGFR wild-type and has a minimally reactive electrophile to reduce off-target labeling. This compound is for research use only and not sold to patients.
  • Inhibition of EGF-stimulated wild type EGFR autophosphorylation expressed in human A549 cells
  • Inhibition of EGFR L858R/T790M double mutant autophosphorylation in human NCI-H1975 cells
  • Excellent selectivity against EGFR wild-type
  • Minimally reactive electrophile reduces off-target labeling