Learn More
Medchemexpress LLC (3S,4S)-Tivantinib | 905854-03-7 | C23H19N3O2 | 1 ML
SDP
Catalog No. 5000359114
Encompass_Preferred
(3S,4S)-Tivantinib is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. It also targets GSK3α and GSK3β, which play an important role in the cellular mechanism of non-small cell lung cancer (NSCLC).
  • Potent and highly selective inhibitor of c-MET
  • Targets GSK3α and GSK3β
  • Relevant for non-small cell lung cancer (NSCLC) research
Catalog No. 50-003-59114 Supplier Medchemexpress LLC Supplier No. HY5068710MM/1ML
(3S,4S)-Tivantinib is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. It also targets GSK3α and GSK3β, which play an important role in the cellular mechanism of non-small cell lung cancer (NSCLC).
  • Potent and highly selective inhibitor of c-MET
  • Targets GSK3α and GSK3β
  • Relevant for non-small cell lung cancer (NSCLC) research