Learn More
Medchemexpress LLC (3S,4S)-Tofacitinib | 1092578-47-6 | MFCD11035920 | 99.8% | 312.37 g/mol | C16H20N6O | 10 MG
SDP
Catalog No. 5000299070
Encompass_Preferred
(3S,4S)-Tofacitinib is the S-enantiomer of the JAK inhibitor tofacitinib and is provided as a research reference compound for biochemical and cellular studies. It is reported to be less active than the clinical racemate and is characterized by CAS 1092578-47-6, formula C16H20N6O, and molecular weight 312.37 g/mol.
  • Enantiomeric reference for JAK signaling studies.
  • High purity (99.75%) suitable for analytical and biochemical assays.
  • Molecular formula C16H20N6O and molecular weight 312.37 g/mol.
  • Supplied as a 10 mg research quantity.
  • Useful as a reference standard for selectivity and potency comparisons.
Catalog No. 50-002-99070 Supplier Medchemexpress LLC Supplier No. HY40354C10MG
(3S,4S)-Tofacitinib is the S-enantiomer of the JAK inhibitor tofacitinib and is provided as a research reference compound for biochemical and cellular studies. It is reported to be less active than the clinical racemate and is characterized by CAS 1092578-47-6, formula C16H20N6O, and molecular weight 312.37 g/mol.
  • Enantiomeric reference for JAK signaling studies.
  • High purity (99.75%) suitable for analytical and biochemical assays.
  • Molecular formula C16H20N6O and molecular weight 312.37 g/mol.
  • Supplied as a 10 mg research quantity.
  • Useful as a reference standard for selectivity and potency comparisons.