Learn More
Medchemexpress LLC (3S,5S)-Atorvastatin | 501121-34-2 | 95.0% | 5 MG
SDP
Catalog No. 5000407681
Encompass_Preferred
(3S,5S)-Atorvastatin is an inactive enantiomer of Atorvastatin. It can activate pregnane X receptor (PXR). Atorvastatin, in its active form, is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids.
  • Increases CYP2B and CYP3A mRNA content with equal ability.
  • Induces luciferase activity with an EC50 of 12.4 μM at 100 μM concentrations.
  • Enhances the intensity of l-tryptophanol fluorescence.
Catalog No. 50-004-07681 Supplier Medchemexpress LLC Supplier No. HYB0589C5MG
(3S,5S)-Atorvastatin is an inactive enantiomer of Atorvastatin. It can activate pregnane X receptor (PXR). Atorvastatin, in its active form, is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids.
  • Increases CYP2B and CYP3A mRNA content with equal ability.
  • Induces luciferase activity with an EC50 of 12.4 μM at 100 μM concentrations.
  • Enhances the intensity of l-tryptophanol fluorescence.