Learn More
Medchemexpress LLC (3S,5S)-atorvastatin | 501121-34-2 | MFCD09839951 | 95.0% | 558.64 g/mol | C33H35FN2O5 | 10 MG
SDP
Catalog No. 5000297353
Encompass_Preferred
(3S,5S)-atorvastatin is the inactive stereoisomer of atorvastatin provided for research use. It is reported to act as a pregnane X receptor (PXR) agonist and is used in pharmacology and metabolic-disease studies. Supplied as a high-purity solid for in vitro and analytical applications.
  • Inactive enantiomer of atorvastatin
  • Acts as pregnane X receptor (PXR) agonist
  • Supplied with reported purity of 95.0%
  • Available in a 10 mg quantity for laboratory use
  • Suitable for in vitro and analytical research; not for human use
Catalog No. 50-002-97353 Supplier Medchemexpress LLC Supplier No. HYB0589C10MG
(3S,5S)-atorvastatin is the inactive stereoisomer of atorvastatin provided for research use. It is reported to act as a pregnane X receptor (PXR) agonist and is used in pharmacology and metabolic-disease studies. Supplied as a high-purity solid for in vitro and analytical applications.
  • Inactive enantiomer of atorvastatin
  • Acts as pregnane X receptor (PXR) agonist
  • Supplied with reported purity of 95.0%
  • Available in a 10 mg quantity for laboratory use
  • Suitable for in vitro and analytical research; not for human use