Learn More
Cayman Chemical (3S,5S)-Atorvastatin (sodium salt), Purity: ≥98%, CAS: 1428118-38-0, 10 mg
SDP
Catalog No. 502965238
Encompass_Preferred
An enantiomer of atorvastatin that has little or no inhibitory activity against HMG-CoA reductase
Catalog No. 50-296-5238 Supplier Cayman Chemical Supplier No. 1846510 MG
An enantiomer of atorvastatin that has little or no inhibitory activity against HMG-CoA reductase