Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3S,5S)-pitavastatin calcium | 254452-92-1 | MFCD28138313 | 99.9% | 440.49 g·mol^-1 | C25H24FNO4 · 1/2 Ca | 5 MG
SDP

Catalog No. 5000223497
Encompass_Preferred

(3S,5S)-Pitavastatin calcium is the 3-epimer of pitavastatin calcium, a potent HMG-CoA reductase inhibitor used for cardiovascular disease research. It is supplied as the calcium salt and is intended for research use, with high reported purity suitable for analytical and biological studies.

  • High purity (99.9%) suitable for analytical and biological assays.
  • Molecular formula C25H24FNO4 · 1/2 Ca.
  • Molecular weight 440.49 g·mol^-1.
  • Available in small milligram quantities for laboratory use.
  • Useful as a reference standard or for impurity profiling.
  • Provided as a solid, white to off-white powder.

Catalog No. 50-002-23497 Supplier Medchemexpress LLC Supplier No. HY1353835MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(3S,5S)-Pitavastatin calcium is the 3-epimer of pitavastatin calcium, a potent HMG-CoA reductase inhibitor used for cardiovascular disease research. It is supplied as the calcium salt and is intended for research use, with high reported purity suitable for analytical and biological studies.

  • High purity (99.9%) suitable for analytical and biological assays.
  • Molecular formula C25H24FNO4 · 1/2 Ca.
  • Molecular weight 440.49 g·mol^-1.
  • Available in small milligram quantities for laboratory use.
  • Useful as a reference standard or for impurity profiling.
  • Provided as a solid, white to off-white powder.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.