Learn More
Echelon Biosciences Research Labs 3S-PTDINS(3,4,5)P3 DIC16 500 ug
SDP
Catalog No. 50720612
Encompass
PtdIns(3,4,5)P3 3-phosphothioate, diC16 (3S-PtdIns(3,4,5)P3, diC16) is a water-soluble, synthetic PI(3,4,5)P3 analog in which the 3-phosphate has been rendered metabolically stable as a phosphorothioate. P-3S908 shows 90% inhibition of PTEN at 0.4M. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-612 Supplier Echelon Biosciences Research Labs Supplier No. P3S916500UG
PtdIns(3,4,5)P3 3-phosphothioate, diC16 (3S-PtdIns(3,4,5)P3, diC16) is a water-soluble, synthetic PI(3,4,5)P3 analog in which the 3-phosphate has been rendered metabolically stable as a phosphorothioate. P-3S908 shows 90% inhibition of PTEN at 0.4M. This item is non returnable