Learn More
Perkin Elmer US LLC 3-Slot Demountable Organics Quartz Torch for Optima 7300 V
Catalog No. 502104733
Encompass_Preferred
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use in organic analysis.
Catalog No. 50-210-4733 Supplier Perkin Elmer US LLC Supplier No. N0771561
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use in organic analysis.