Learn More
Perkin Elmer US LLC 3-Slot Demountable Quartz Torch for Optima 8x00
Catalog No. 502112265
Encompass_Preferred
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use in organic analysis. It is included in the Organics Sample Introduction Kit for Optima 8x00 (N0780608).
Catalog No. 50-211-2265 Supplier Perkin Elmer US LLC Supplier No. N0780132
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use in organic analysis. It is included in the Organics Sample Introduction Kit for Optima 8x00 (N0780608).