Learn More
Cole-Parmer PIPET-AID Drummond Portable XP, 10 mL/sec, 110 VAC
Catalog No. NC9796403 Shop All Cole-Parmer Products
Encompass_Preferred
Three speed range options for more precise control. Set the fill and empty speeds independently to fast, medium, or slow. Additional speed control is provided depending on how far each button is depressed. Features a built-in pump—there are no power lines or tubing to drag around. Measures only 7" high, making it ideal for use in the lab or field. Bodies are constructed of Delrin plastic. Removable nosepiece is autoclavable. Operates on a rechargeable nickel metal hydride (NiMH) battery. Can be used continuously for two hours before recharging is required.
Catalog No. NC9796403 Supplier Cole-Parmer Supplier No. 0789740
Three speed range options for more precise control. Set the fill and empty speeds independently to fast, medium, or slow. Additional speed control is provided depending on how far each button is depressed. Features a built-in pump—there are no power lines or tubing to drag around. Measures only 7" high, making it ideal for use in the lab or field. Bodies are constructed of Delrin plastic. Removable nosepiece is autoclavable. Operates on a rechargeable nickel metal hydride (NiMH) battery. Can be used continuously for two hours before recharging is required.