Learn More
Abcam 3T3-L1 Differentiation Kit, 100ML
Catalog No. 502205913
Encompass_Preferred
This 3T3-L1 Differentiation Kit provides enough supplements to make 100 ml of differentiation medium and 600 ml of maintenance medium which is sufficient material for 12 100 mm culture dishes. The Differentiation Cocktail provides a final concentration of 1.5 μg/ml insulin insulin.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5913 Supplier Abcam Supplier No. AB287843100ML
This 3T3-L1 Differentiation Kit provides enough supplements to make 100 ml of differentiation medium and 600 ml of maintenance medium which is sufficient material for 12 100 mm culture dishes. The Differentiation Cocktail provides a final concentration of 1.5 μg/ml insulin insulin.
The product is subject to the following: Abcam Restricted Use Statement