Learn More
ATCC 3T3-L1; Embryonic Fibroblast; Mouse
Catalog No. 502384023
Encompass
3T3-L1 is a fibroblast that was isolated from the embryo of a mouse. This cell line can be used to study the basic cellular mechanisms associated with diabetes, obesity, and related disorders.
Catalog No. 50-238-4023 Supplier ATCC Supplier No. CL173
3T3-L1 is a fibroblast that was isolated from the embryo of a mouse. This cell line can be used to study the basic cellular mechanisms associated with diabetes, obesity, and related disorders.