Learn More
Cayman Chemical 3-thio-Pheneacrylic Acid methyl ester, Purity: ≥98%, CAS: 135835-43-7, 1 g
SDP
Catalog No. 502888054
Encompass_Preferred
A synthetic intermediate useful for pharmaceutical synthesis
Catalog No. 50-288-8054 Supplier Cayman Chemical Supplier No. 100071021 G
A synthetic intermediate useful for pharmaceutical synthesis