Learn More
Andwin Scientific DURACELL Lithium Batteries 3V, 20 dia. x 2.5mm H 150mA hours, 6pk
SDP
Catalog No. NC9650466
Encompass_Preferred
DURACELL Lithium Batteries, For automobile security, electronic organizers. 3V, 20 dia. x 2.5mm H 150mA hours. Batteries contain lithium-manganese dioxide. With wide operating temperature range of -41degreeC to 60degreeC
Catalog No. NC9650466 Supplier Andwin Scientific Supplier No. 13709940
DURACELL Lithium Batteries, For automobile security, electronic organizers. 3V, 20 dia. x 2.5mm H 150mA hours. Batteries contain lithium-manganese dioxide. With wide operating temperature range of -41degreeC to 60degreeC