Learn More
New Jersey Fluid Systems LLC 3WAY SS VALVE
Catalog No. NC9635859
Encompass
316 Stainless Steel Swagelok 3-Way Ball Valve. Panel Mount. 1/4 in. OD Tube Fitting ends. -65F to 300F 2,500psi max. PTFE packing; Cv=0.35
This item is not returnable. View return policy
Catalog No. NC9635859 Supplier New Jersey Fluid Systems LLC Supplier No. SS42GXS4
316 Stainless Steel Swagelok 3-Way Ball Valve. Panel Mount. 1/4 in. OD Tube Fitting ends. -65F to 300F 2,500psi max. PTFE packing; Cv=0.35