Learn More
Active Motif Sortag-IT™ 3X DBCO Click Labeling Kit, 3 x 100 µg
SDP
Catalog No. NC3665915
Encompass
Sortag-IT™ Labeling Kits are designed for site-specific labeling of Active Motif's highly specific AbFlex® recombinant antibodies (rAb) via the Sortase tag recognition sequence (LPXTG) that is incorporated into the heavy chains of each AbFlex antibody. Directly conjugate fluorophores, enzymes (HRP, AP), biotin, peptides, DNA, carbohydrates, or other labels to the terminal end of the heavy chain to ensure the antigen binding site remains available for target recognition.
Catalog No. NC3665915 Supplier Active Motif Supplier No. 13116
Sortag-IT™ Labeling Kits are designed for site-specific labeling of Active Motif's highly specific AbFlex® recombinant antibodies (rAb) via the Sortase tag recognition sequence (LPXTG) that is incorporated into the heavy chains of each AbFlex antibody. Directly conjugate fluorophores, enzymes (HRP, AP), biotin, peptides, DNA, carbohydrates, or other labels to the terminal end of the heavy chain to ensure the antigen binding site remains available for target recognition.