Learn More
Apexbio Technology LLC 3X (DYKDDDDK) Peptide, 4mg. Cas: MFCD: NA. Synthetic peptide tag
SDP
Supplier:  Apexbio Technology LLC A60014
Encompass_Preferred
The FLAG-tag system utilizes a short, hydrophilic 8- amino-acid peptide that is fused to the protein of interest. The FLAG peptide binds to the antibody M1. Whether binding is calcium-dependent manner or -independent remains controversial
Catalog No. NC0792928