Learn More
Medchemexpress LLC HY-P0319 5mg , 3X FLAG peptide CAS:402750-12-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP03195MG
Encompass_Preferred
HY-P0319 5mg 3X FLAG peptide CAS: 402750-12-3 3X FLAG Peptide is a synthetic peptide with a 3-time repeated DYKXXD motif. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9842