Learn More
Medchemexpress LLC 3X FLAG peptide TFA, 10 mg
SDP
Catalog No. NC1732472
Encompass_Preferred
3X FLAG peptide TFA is a synthetic peptide with a 3-time repeated DYKXXD motif.
Catalog No. NC1732472 Supplier Medchemexpress LLC Supplier No. HYP0319A10MG
Add to Cart
Add to Cart
3X FLAG peptide TFA is a synthetic peptide with a 3-time repeated DYKXXD motif.