Learn More
Medchemexpress LLC HY-P0319A 25mg Medchemexpress, 3X FLAG peptide (TFA) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0319A25MG
Encompass_Preferred
Medchemexpress, HY-P0319A 25mg 3X FLAG peptide (TFA) CAS:0 3X FLAG peptide TFA is a synthetic peptide with a 3-time repeated DYKXXD motif. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9433