Learn More
Bachem 3X Flag Peptide Trifluoroacetate
SDP
Supplier:  Bachem 4046200.0005
Encompass_Preferred
3X Flag Peptide Trifluoroacetate, H-Met-Asp-Tyr-Lys-Asp-His-Asp-Gly-Asp-Tyr-Lys-Asp-His-Asp-Ile-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys-OH, 402750-12-3, C120H169N31O49S, Mr 2861.91, 5mg. Triple FLAG.
Catalog No. NC1139866