Learn More
Uline Cleanroom Labels - 3 x 1"
Catalog No. NC3261499
Encompass
This item must be ordered in quantities of 4 each per supplier requirements.
Label beakers, bags and samples with no risk of contamination. Matte polypropylene labels with acrylic adhesive. Plastic core won't shed particles.
Catalog No. NC3261499 Supplier Uline Supplier No. S25228
Add to Cart
Add to Cart
This item must be ordered in quantities of 4 each per supplier requirements.
Label beakers, bags and samples with no risk of contamination. Matte polypropylene labels with acrylic adhesive. Plastic core won't shed particles.