Learn More
Starna Cells Inc 3X3X45 UV QUARTZ CUVET/STOP
SDP
Catalog No. NC9907273
Encompass_Preferred
Square Micro Fluorometer Cell, Stoppered,Window material: Spectrosil Quartz or equivalent,useable range: 170 to 2700nm,Path Length:3 mm,Exterior (W x L x H): 5.5 x 5.5 x 48 mm, Interior (W x L): 3 x 3 mm, Nominal Volume: 0.315 ml, Number Po
Catalog No. NC9907273 Supplier Starna Cells Inc Supplier No. 233.45Q3
Square Micro Fluorometer Cell, Stoppered,Window material: Spectrosil Quartz or equivalent,useable range: 170 to 2700nm,Path Length:3 mm,Exterior (W x L x H): 5.5 x 5.5 x 48 mm, Interior (W x L): 3 x 3 mm, Nominal Volume: 0.315 ml, Number Po