Learn More
Supply Source 3"W x 5"L Reclosable Poly Bag, 2.0 Mil, 1000/Carton
Catalog No. NC1808357
Encompass_Preferred
reclosable 2-mil poly bag measuring 3" x 5" has a pinch track zipper closure that aids to keep products clean and moisture free.The Reclosable 2-mil poly bag measuring 3" x 5" is made from 100 percent virgin high clarity polyethylene film for added durability. Bag meets all requirements of FDA and USDA specifications for food contact.Overall Dimensions: 3"(W) x 5"(L)2 MilsMeets FDA/USDA SpecificationsSide weldNo need to tie, tape or heat seal
This item is not returnable. View return policy
Catalog No. NC1808357 Supplier Supply Source Supplier No. 3550A100PK
reclosable 2-mil poly bag measuring 3" x 5" has a pinch track zipper closure that aids to keep products clean and moisture free.The Reclosable 2-mil poly bag measuring 3" x 5" is made from 100 percent virgin high clarity polyethylene film for added durability. Bag meets all requirements of FDA and USDA specifications for food contact.Overall Dimensions: 3"(W) x 5"(L)2 MilsMeets FDA/USDA SpecificationsSide weldNo need to tie, tape or heat seal