Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3Z)-2-amino-4-(4-hydroxyphenyl)buta-1,3-diene-1,1,3-tricarbonitrile | 144978-82-5 | 98.6% | 236.23 g/mol | C13H8N4O | 5 MG
SDP

Catalog No. 5000263245
Encompass_Preferred

Tyrphostin AG 112 is a small-molecule inhibitor of epidermal growth factor receptor (EGFR) phosphorylation used in biochemical and cellular research. The compound is supplied as a solid with high purity and is compatible with DMSO-based solutions for in vitro testing and with co-solvent formulations for in vivo studies.

  • Molecular formula: C13H8N4O.
  • Molecular weight: 236.23 g/mol.
  • Purity: 98.6%.
  • Appearance: solid.
  • Solubility: DMSO soluble (125 mg/mL); in vivo co-solvent formulations give clear solutions.
  • Available pack sizes: 5 mg, 10 mg, 25 mg, larger sizes by quote.

Catalog No. 50-002-63245 Supplier Medchemexpress LLC Supplier No. HY1124745MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrphostin AG 112 is a small-molecule inhibitor of epidermal growth factor receptor (EGFR) phosphorylation used in biochemical and cellular research. The compound is supplied as a solid with high purity and is compatible with DMSO-based solutions for in vitro testing and with co-solvent formulations for in vivo studies.

  • Molecular formula: C13H8N4O.
  • Molecular weight: 236.23 g/mol.
  • Purity: 98.6%.
  • Appearance: solid.
  • Solubility: DMSO soluble (125 mg/mL); in vivo co-solvent formulations give clear solutions.
  • Available pack sizes: 5 mg, 10 mg, 25 mg, larger sizes by quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.