Learn More
Medchemexpress LLC (3Z)-3-(2-pyridinylmethylene)-1,3-dihydro-2H-indol-2-one | 187325-53-7 | MFCD00435358 | 98.0% | 222.25 | C14H10N2O | 10MG
SDP
Catalog No. 5000234972
Encompass_Preferred
GSK-3β inhibitor 1 is a potent small-molecule inhibitor of glycogen synthase kinase-3β (compound 3a) with an enzymatic IC50 of 4.19 nM and demonstrated antidiabetic efficacy in streptozotocin-treated obese rat models. The compound is provided as a high-purity research chemical (98.04%) in solid form and as DMSO solutions, and is suitable for in vitro and in vivo research applications.
  • Potent enzymatic inhibition (IC50 = 4.19 nM).
  • Demonstrated antidiabetic efficacy in animal models.
  • High purity suitable for research applications.
  • Available as solid and DMSO solution formats.
  • Supplied in multiple sizes for scalable experiments.
  • Molecular weight 222.25 g·mol⁻¹; formula C14H10N2O.
Catalog No. 50-002-34972 Supplier Medchemexpress LLC Supplier No. HY12614410MG
GSK-3β inhibitor 1 is a potent small-molecule inhibitor of glycogen synthase kinase-3β (compound 3a) with an enzymatic IC50 of 4.19 nM and demonstrated antidiabetic efficacy in streptozotocin-treated obese rat models. The compound is provided as a high-purity research chemical (98.04%) in solid form and as DMSO solutions, and is suitable for in vitro and in vivo research applications.
  • Potent enzymatic inhibition (IC50 = 4.19 nM).
  • Demonstrated antidiabetic efficacy in animal models.
  • High purity suitable for research applications.
  • Available as solid and DMSO solution formats.
  • Supplied in multiple sizes for scalable experiments.
  • Molecular weight 222.25 g·mol⁻¹; formula C14H10N2O.