Learn More
Roboz Surgical 4-0 GUT CHROMIC 18IN CE-3
SDP
Catalog No. NC9615255
Encompass
Suture Material With C-12 Needle,Sterile, 18 in. Length, Box Of 36. Size: 4-0; Gut, Chromic, Absorbable
Catalog No. NC9615255 Supplier Roboz Surgical Supplier No. SUT57031
Add to Cart
Add to Cart
Suture Material With C-12 Needle,Sterile, 18 in. Length, Box Of 36. Size: 4-0; Gut, Chromic, Absorbable