Learn More
Perkin Elmer US LLC 4.0 mL Sample Loop, 1 mm ID, connect to ports #1 and #4
Catalog No. 502105232
Encompass_Preferred
Multipurpose PFA FAST loops for high-flow valves. Recommended for use with 1.0 mm i.d. (gray) probe. 1.6 mm o.d., 1/4-28 fittings.
Catalog No. 50-210-5232 Supplier Perkin Elmer US LLC Supplier No. N0777480
Multipurpose PFA FAST loops for high-flow valves. Recommended for use with 1.0 mm i.d. (gray) probe. 1.6 mm o.d., 1/4-28 fittings.