Learn More
Braintree Scientific 4-0 Silk Suture, 1/2 circle tappered, double
SDP
Catalog No. 501955976
Encompass
AT-2 needle, tapered pt. 1/2 circle, wire dia. 0.0180, channel - min. 0.0217 min/max 0.0237, length 0.0687" 17.4mm, 4-0, 24" **Sold for research purposes only and not for human use**
Catalog No. 50-195-5976 Supplier Braintree Scientific Supplier No. SUT7116
AT-2 needle, tapered pt. 1/2 circle, wire dia. 0.0180, channel - min. 0.0217 min/max 0.0237, length 0.0687" 17.4mm, 4-0, 24" **Sold for research purposes only and not for human use**