Learn More
Pipette Com Drummond Pipet-Aid Portable, 110V Charger Included
SDP
Catalog No. NC1742482
Encompass
The Original Portable Pipet-Aid Pipette Controller brings reliability and durability into the laboratory with a precision device that has become a staple for millions of technicians across the world. As convenient as it is powerful, the Original Portable PIpet-Aid Pipette Controller provides cordless usability and proven technology. The flow rate is better than 25 mL at 5 seconds with 100mL pipette and the filtered, failsafe nosepiece eliminates cross-contamination due to over-pipetting. See why the Original Portable Pipet-Aid Pipette Controller is the industry standard that changed the industry.
Catalog No. NC1742482 Supplier Pipette Com Supplier No. DP100
The Original Portable Pipet-Aid Pipette Controller brings reliability and durability into the laboratory with a precision device that has become a staple for millions of technicians across the world. As convenient as it is powerful, the Original Portable PIpet-Aid Pipette Controller provides cordless usability and proven technology. The flow rate is better than 25 mL at 5 seconds with 100mL pipette and the filtered, failsafe nosepiece eliminates cross-contamination due to over-pipetting. See why the Original Portable Pipet-Aid Pipette Controller is the industry standard that changed the industry.