Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1,1,3,3-Tetramethylbutyl)phenol 95.0+%, TCI America™
SDP

Catalog No. T0144500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0144500G 500 g
T014425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0144500G Supplier TCI America Supplier No. T0144500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 140-66-9
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.329
MDL Number MFCD00002368
InChI Key ISAVYTVYFVQUDY-UHFFFAOYSA-N
Synonym 4-tert-octylphenol, p-tert-octylphenol, 4-t-octylphenol, 4-1,1,3,3-tetramethylbutyl phenol, 4-2,4,4-trimethylpentan-2-yl phenol, p-terc.oktylfenol, para-tert-octylphenol, p-1,1,3,3-tetramethylbutyl phenol, phenol, 4-1,1,3,3-tetramethylbutyl, phenol, p-tert-octyl
PubChem CID 8814
ChEBI CHEBI:34445
IUPAC Name 4-(2,4,4-trimethylpentan-2-yl)phenol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)O

Specifications

Melting Point 83°C
Color White
Boiling Point 279°C
Quantity 500 g
UN Number 3077
Formula Weight 206.33
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1,1,3,3-Tetramethylbutyl)phenol
EINECSNumber (3)-0503
RTECSNumber SM9625000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.