Learn More
Medchemexpress LLC 4-((((1-(2-(2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethoxy)ethyl)-1h-1, | 98.4% | 795.72 | 5 MG
SDP
Catalog No. 5000384982
Encompass_Preferred
TO-1187 TFA is a selective HDAC6 PROTAC degrader with a DC50 of 5.81 nM. It promotes the ubiquitination and degradation of HDAC6 and is used in the study of hematological malignancies and solid tumors. The compound is composed of an HDAC6 ligand, a CRBN ligase ligand, and a linker.
  • Selective PROTAC degrader
  • Promotes ubiquitination and degradation of HDAC6
  • Applicable for studying hematological malignancies and solid tumors
Catalog No. 50-003-84982 Supplier Medchemexpress LLC Supplier No. HY173266A5MG
TO-1187 TFA is a selective HDAC6 PROTAC degrader with a DC50 of 5.81 nM. It promotes the ubiquitination and degradation of HDAC6 and is used in the study of hematological malignancies and solid tumors. The compound is composed of an HDAC6 ligand, a CRBN ligase ligand, and a linker.
  • Selective PROTAC degrader
  • Promotes ubiquitination and degradation of HDAC6
  • Applicable for studying hematological malignancies and solid tumors