Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (4-(1,2,4,5-Tetrazin-3-Yl)Phenyl)Methanamine Hydrochloride

Catalog No. 76159125MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-159-125MG 25 mg
76-159-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 76-159-125MG Supplier Sigma Aldrich Supplier No. 76159125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(4-(1,2,4,5-Tetrazin-3-Yl)Phenyl)Methanamine Hydrochloride

Specifications

CAS 1345866-68-3
Melting Point 206°C to 225°C (decomposition)
Molecular Formula C9H9N5 -+ xHCl
Linear Formula C9H9N5 -+ xHCl
MDL Number MFCD23097050
Quantity 25 mg
Synonym Benzylamino tetrazine hydrochloride; H-Tz-Bz-NH3Cl hydrochloride
Molecular Weight (g/mol) 187.20 (free base basis) g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.