Learn More
Medchemexpress LLC 4-[1-(2-fluoropyridin-3-yl)-5-methyl-1H-1,2,3-triazol-4-yl]-N-isopropyl-N-methyl-3,6-dihydropyridine-... | 873551-53-2 | 99.3% | 358.41 g/mol | C18H23FN6O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10040510MG
Encompass_Preferred
FTIDC is a selective, noncompetitive allosteric antagonist of metabotropic glutamate receptor 1 (mGluR1) intended for research use. It is orally active and demonstrates potent antagonism against recombinant mGluR1, supporting pharmacology and neuroscience studies.
  • Potent mGluR1 antagonism (IC50 5.8 nM for human mGluR1a).
  • Selective noncompetitive allosteric mechanism.
  • High chemical purity suitable for research applications.
  • Defined physicochemical properties: C18H23FN6O; MW 358.41 g/mol; CAS 873551-53-2.
  • Supplied in small-scale quantities appropriate for laboratory experiments.
Catalog No. 50-002-14055