Learn More
Sklar Instruments Single Use, Econo-Sterile, Iris Scissors, Sklar 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, Sterile, CS of 100.
SDP
Catalog No. 501094328 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, Sterile, CS of 100. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal.
Catalog No. 50-109-4328 Supplier Sklar Instruments Supplier No. 962651
Single Use, Econo-Sterile, Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, Sterile, CS of 100. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal.