Learn More
Sklar Instruments Scissors, Supercut Sklar Iris Scissors, Sklar 4-1/2 in., Sharp/Sharp, Curved, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Stainless Steel. EA.
SDP
Catalog No. 501093451 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Supercut Sklarhone Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Stainless Steel. EA. Supercut Sklarhone Iris scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone scissors minimize tearing of tissue so cuts and edges are smooth and clean. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. This product is straight with one razor edge blade, one micro-serrated edge blade, sharp tips, and an overall length of 4-1/2 inches.
Catalog No. 50-109-3451 Supplier Sklar Instruments Supplier No. 153457
Scissors, Supercut Sklarhone Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Stainless Steel. EA. Supercut Sklarhone Iris scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone scissors minimize tearing of tissue so cuts and edges are smooth and clean. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. This product is straight with one razor edge blade, one micro-serrated edge blade, sharp tips, and an overall length of 4-1/2 inches.