Learn More
Sklar Instruments Scissors, Sklar Operating Scissors, Sklar 4-1/2 in., Sharp/Sharp, Curved, Smooth, Extra Light, Stainless Steel, Reusable, EA.
SDP
Catalog No. 501093667 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Sklarlite Operating Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Extra Light, Stainless Steel, Reusable, EA. Sklarlite Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Sklarlite instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability.
Catalog No. 50-109-3667 Supplier Sklar Instruments Supplier No. 231124
Scissors, Sklarlite Operating Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Extra Light, Stainless Steel, Reusable, EA. Sklarlite Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Sklarlite instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability.